missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Stainless Steel spheres, 3mm (0.12 in.) dia., Type 302

Brand:  Alfa Aesar™ 042017.KM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 25 pieces

 View more versions of this product

Product Code. 11365949

  • £16.54 / Pack of 25


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

Stainless Steel spheres, Type 302
3mm (0.12 in.) dia.
MFCD00148499

Specifications

Fe:Cr:Ni:Mn:Si; 70:18:9:2:1 wt%
25 pieces
MFCD00148499
Stainless Steel spheres, Type 302
3mm (0.12 in.) dia.
Videos
SDS