missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Stainless Steel spheres, 3mm (0.12 in.) dia., Type 302

Brand:  Thermo Scientific Alfa Aesar 042017.KN

Stainless Steel spheres, 3mm (0.12 in.) dia., Type 302

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11375949

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.