missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel Spheres, 2mm (0.08 in ) dia., Alfa Aesar™

Brand:  Alfa Aesar 042019.KM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 25 Pieces

 View more versions of this product

Product Code. 11305959

  • £16.70 / Pack of 25


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Stainless Steel
Spheres, 2mm (0.08 In ) dia.
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
MFCD00148499

Specifications

Stainless Steel
Spheres
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
25 Pieces
Spheres, 2mm (0.08 In ) dia.
MFCD00148499
Videos
SDS