missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Stainless Steel Sliding Shelf Rack

The Stainless Steel Sliding Shelf Rack is a comfort rack for upright freezers that allows a more ergonomic operating process when working with your samples. They are made of stainless steel and can be used with all freezer models.

Brand:  Tenak™ TE25084

Additional Details : Weight : 2.20000kg

 View more versions of this product

Product Code. 15131289

  • £36.20 / Each
Estimated Shipment: 09-08-2024
to see stock.
Add to basket

Description

Description

  • Stainless steel
  • Fit all Upright freezer models
Specifications

Specifications

Sliding Shelf Rack
Boxes
5
1 Ea.
130 mm
1 x 5
Stainless Steel
Upright Freezers
140 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers