missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Stainless Steel rod, 9.5mm (0.375 in.) dia., Type 303

Brand:  Alfa Aesar™ 042707.RM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 90cm

 View more versions of this product

Product Code. 11355467

  • £37.74 / 90cm


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Specifications

Chemical Identifiers

Stainless Steel rod, Type 303
9.5mm (0.375 in.) dia.
MFCD00148499

Specifications

Fe:Cr:Ni; 73:18:9 wt%
90cm
Rod
Stainless Steel rod, Type 303
9.5mm (0.375 in.) dia.
MFCD00148499
Videos
SDS