missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Stainless Steel rod, 9.5mm (0.375 in.) dia., Type 303

Brand:  Thermo Scientific Alfa Aesar 042707.BV

Stainless Steel rod, 9.5mm (0.375 in.) dia., Type 303

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11365467

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.