Learn More
Endecotts™ Stainless Steel Receiver
Full height receivers for use with test sieves. Endecotts™ Stainless Steel Receivers are widely used with test sieves for all types of laboratory sampling and particle size analysis.
Brand: Endecotts™ 300S/STREC
Code : 03
Additional Details : Weight : 0.50000kg
Description
Full height receivers for use with test sieves. Endecotts™ Stainless Steel Receivers are widely used with test sieves for all types of laboratory sampling and particle size analysis.
- Stainless steel construction
- For use with Stainless Steel Woven Wire Mesh Test Sieves
Specifications
Receiver | |
300 mm | |
1 Ea. |
Stainless Steel | |
For sieves |