Learn More
Stainless Steel powder, -40+80 mesh, Type 316, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 042944.30
43.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.25000kg
Description
Stainless Steel powder, Type 316 is a increased resistance to chemical corrosion for the addition of Molybdenum. Also used in a wide variety of home and commercial applications, including, chemical process equipment, food and beverage industry and fasteners.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel powder, Type 316 | |
MFCD00148499 | |
Insoluble in water. | |
Powder |
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% | |
250 g | |
−40+80 Mesh |
RUO – Research Use Only