Learn More
Stainless Steel powder, -325 mesh, Type 316-L, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 088390.36
86.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.50000kg
Description
Stainless Steel powder is employed in pharmaceuticals, marine applications, architectural applications, medical implants, including pins, screws and orthopaedic implants like total hip and knee replacements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gray | |
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% | |
500 g | |
−325 Mesh |
Stainless Steel powder, Type 316-L | |
MFCD00148499 | |
Insoluble in water. | |
Powder |
RUO – Research Use Only