Learn More
Stainless Steel powder, -140 mesh, Type 303-L, Thermo Scientific Chemicals
£64.20 - £222.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316098
|
Thermo Scientific Alfa Aesar
011084.A1 |
1 kg |
£222.00
1kg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15450997
|
Thermo Scientific Alfa Aesar
011084.30 |
250 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This offers moderate corrosion resistance. Alloy 303 is frequently used in applications that require parts to be heavily machined. It is used in nuts and bolts, aircraft fittings, gears and screws.