Learn More
Stainless Steel powder, -100 mesh, Type 304-L, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 011085.36
Code : NEW
Additional Details : Weight : 0.50000kg
Description
Stainless Steel powder, -100 mesh, Type 304-L is the most common stainless steel alloy widely used in wire weaving. It has excellent corrosion resistance and can withstand temperatures of 1100 degrees F. It has better welding characteristics for lower carbon content.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel powder, Type 304-L | |
MFCD00148499 | |
Insoluble in water. | |
Powder |
Fe:Cr:Ni; 70:19:11 wt% | |
500 g | |
−100 Mesh |
RUO – Research Use Only