Learn More
DWK Life Sciences Plate Holder, for glass ceramic laboratory protection plate
Ideal holder for glass ceramic plates for mounting on tripods.
Brand:  DWK Life Sciences 290785809
Code : NEW
Additional Details : Weight : 1.16500kg
 View more versions of this product
Product Code. 11772909
  • £221.00 / Pack of 5
Estimated Shipment: 24-07-2024
to see stock.
Add to basket
Description
Description
In the laboratory, solutions are often heated. This step can be carried out using different techniques, e.g. in a water or sand bath, with a heating hood or classically using a Bunsen burner. If the Bunsen burner is used as the heat source, it is recommended to use a tripod or square foot and a corresponding glass ceramic plate. The vessel to be heated is placed on the glass ceramic plate with the Bunsen burner underneath. Contamination of the glass ceramic plate can be removed with the cleaning scraper.
  • Made from heat-resistant chrome-nickel steel with
Specifications
Specifications
Chrome-nickel Steel
175 mm
Plate holder for glass ceramic plates; Heating; Suitable for mounting on tripods
5/Pk.