Fisherbrand™ Stainless Steel OIML Class M1 Calibration Weight, ISO 17025 Certified (UKAS equivalent)
Overview
Brand: Fisherbrand™
UNSPSC: 41111505
Code: 42
Additional Details:
Additional Details: Weight: 2.00000kg
Product Code. 12901421
 N/A
N/A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Specification
Case Material Wooden
The Fisherbrand™ range offers highly accurate individual weights and weight sets, with nominal mass values from 1mg to 10kg. Both cylindrical and leaf weights are available. With the Fisherbrand™ range including both non-certified and certified cylindrical calibration weights that conform to OIML class F1, F2, E2 and M1 specifications. E2 leaf weights are available too, either as non-certified or certified. Sets of weights are also available within this range, as either certified or non-certified.
Alternative Products
Documentation
Technical documents