Learn More
Fisherbrand™ Stainless Steel OIML Class F1 Calibration Weight Sets With ISO 17025 Certified (UKAS equivalent)
Brand:  Fisherbrand™
Code : 42
Additional Details : Weight : 0.00100kg
Product Code. 12981341
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
The Fisherbrand™ range offers highly accurate individual weights and weight sets, with nominal mass values from 1mg to 10kg. Both cylindrical and leaf weights are available. With the Fisherbrand™ range including both non-certified and certified cylindrical calibration weights that conform to OIML class F1, F2, E2 and M1 specifications. E2 leaf weights are available too, either as non-certified or certified. Sets of weights are also available within this range, as either certified or non-certified.
Specifications
Specifications
Calibration Weight Set
OIML Class F1
2 to 500mg
Leaf Weight
2 to 500mg
1Set
Stainless Steel
ISO 17025 Certified (UKAS equivalent)
Videos
Documentation
Technical Documents