Learn More
Fisherbrand™ Stainless Steel E2 Leaf Calibration Weight
Ensure precise calibration and comparative measurements. Fisherbrand™ Stainless Steel Leaf E2 Calibration Weight is a highly accurate individual weight with a nominal mass value of 5 or 10mg. These leaf weights conform to class E2 requirements and are ISO 17025 certified.
Brand:  Fisherbrand™
Code : 42
Additional Details : Weight : 0.00010kg
Product Code. 12971321
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
Leaf weights for use as mass standards and standard weights are constructed of durable stainless steel, and are available with ISO 17025 certificate for use in testing and calibration labs
  • Available in a range of nine nominal weights (1, 2, 5, 10, 20, 50, 100, 200 and 500mg)
  • For quality assurance in accordance with ISO 9001ff
    • Adjustment, testing and calibration instruments for class I precision scales
    • High-precision weighing or making comparative measurements on class I scales or measurement comparators
    • Quality assurance of scales with fewer than 1,000,000 steps
    • Weight for calibration and official calibration of weights of classes F1 and F2
Specifications
Specifications
Leaf Calibration Weight
Stainless Steel
5mg
E2
Wooden
Videos
Documentation
Technical Documents