missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Stainless Steel Syringe Replacement Needles

Replace needles on syringes with removable needles using Use B Braun™ Sterican™ Stainless Steel Syringe Replacement Needles. for relevant routine, pre- and post-op patient needs.

Brand:  B Braun™ 4657624

Code : Z1

Additional Details : Weight : 0.10800kg

 View more versions of this product

Product Code. 10635913

  • £17.02 / Pack of 100
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sterican™
  • Blunt
  • Stainless steel
  • Latex-free
  • Sterile
Specifications

Specifications

Stainless Steel
0.7 mm
30 mm
Sterile
22 G
Yes
Needle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers