Learn More
Statebourne Cryogenics™ CryoStor™ Cryogenic Cylinders
Helps facilitate the transfer of cryogenic liquid from the CryoStor™ supply vessels. Statebourne Cryogenics™ CryoStor Cryogenic Cylinders are suitable for the supply of liquid or gaseous nitrogen and argon.
Brand:  Statebourne Cryogenics™ 9911075
Code : 04
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11934235
  • £6571.86 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Built to ASME VIII Div. 1 or BS 5500 standards
  • Stainless steel construction
  • All models have castors for easy, convenient handling
  • Dual fill/decant outlets standard on all head manifolds
  • Standard pressure building circuit with regulator
  • With contents gauge indicator
  • Colour-coded liquid and gas delivery valves for easy identification
  • Towing bars to assist easy handling
Specifications
Specifications
Stainless Steel
1378mm
Contents gauge indicator
ASME VIII Div. 1 or BS 5500 standards
180L
720 dia.mm
Stainless Steel
0.9%/day
Cell storage systems
Suitable for the Supply of Liquid or Gaseous Nitrogen and Argon
Yes
CryoStor 180
269kg
640mm
Low Pressure Cryogenic LN2 Cylinder
Tow -bar handles, automatic pressure build circuit, color-coded and labelled valves, twin liquid fill/decant valve, electronic contents indicator
640 dia. x 1378mmH
124kg
3 Years
Videos
Documentation
Technical Documents
View accessory products