missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Stainless Steel Injection Needle

Brand:  Socorex™ 371.1210UN

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15203168

  • £1.60 / Each


Estimated Shipment
29-03-2018
Add to basket

Specifications

Specifications

Yes
1.2mm
Stainless steel
18 ga.
18 ga. x 3/8 in.
10mm
Injection Needle
Videos