Learn More
Fritsch™ Stainless Steel Grinding Balls
Brand: Fritsch™ 55.0200.10
Code : Z3
Additional Details : Weight : 1.00000kg
Specifications
Grinding Balls | |
20 mm | |
Planetary Mono Mill Pulverisette 6 Classic Line |