Learn More
Stainless Steel gauze, 500 mesh woven from 0.02mm (0.0009in) dia wire, Type 304, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 045140.RF
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel gauze, Type 304 | |
Fe:Cr:Ni; 70:19:11 wt% | |
30 x 30 cm |
500 mesh woven from 0.02mm (0.0009 in.) dia. wire | |
MFCD00148499 | |
500 Mesh |
RUO – Research Use Only