missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Stainless Steel gauze, 400 mesh woven from 0.028mm (0.0011 in.) dia. wire, twilled weave, Type 304

Brand:  Alfa Aesar™ 042012.VA

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 15 x 15cm

 View more versions of this product

Product Code. 11375037

  • £100.60 / Each


Estimated Shipment date
from Supplier 28-02-2018
Add to basket

Specifications

Chemical Identifiers

Stainless Steel gauze, Type 304
Woven from 0.028mm (0.0011 in.) dia. wire
MFCD00148499

Specifications

Fe:Cr:Ni; 70:19:11 wt%
400 mesh
Woven from 0.028mm (0.0011 in.) dia. wire
Twilled Weave Wire Cloth
Stainless Steel gauze, Type 304
15 x 15cm
Odorless
MFCD00148499
Videos
SDS