missing translation for 'onlineSavingsMsg'
Learn More

Stainless-Steel foil, Type 304, Alfa Aesar™

Brand:  Alfa Aesar 041583.FI

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 50 x 50mm

 View more versions of this product

Product Code. 11480401

  • £26.20 / Each


Estimated Shipment date
from Supplier 24-01-2018
Add to basket

Specifications

Chemical Identifiers

Stainless Steel
Foil, 0.1mm (0.004 in.) thick
Fe:Cr:Ni; 70:19:11 wt%
MFCD00148499

Specifications

Stainless Steel
50 x 50mm
Odorless
Fe:Cr:Ni; 70:19:11 wt%
Silver
Silver -colored foil
Foil, 0.1mm (0.004 in.) thick
MFCD00148499
Videos
SDS