Learn More
Stainless Steel foil, 0.5mm (0.02in) thick, Type 304, Thermo Scientific Chemicals
£23.50 - £49.50
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11348708
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
041585.FF
25 x 25 mm
£23.50
25mm
View alternative products
Save Up to 
»
Estimated Shipment: 23-04-2024
to see stock.
11400411
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
041585.FI
50 x 50 mm
£34.20
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
11338708
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
041585.GH
100 x 100 mm
£49.50
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Description
Description
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.