missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel Foil, 0.25mm (0.01 in.) thick, Alfa Aesar™

Brand:  Alfa Aesar 041584.FF

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 25 x 25mm

 View more versions of this product

Product Code. 11318708

  • £17.30 / 25mm


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

Stainless Steel
Foil, 0.25mm (0.01 in.) thick
Fe:Cr:Ni; 70:19:11 wt%
MFCD00148499

Specifications

Stainless Steel
25 x 25mm
Odorless
Fe:Cr:Ni; 70:19:11 wt%
Silver
Silver -colored foil
Foil, 0.25mm (0.01 in.) thick
MFCD00148499
Videos
SDS