Learn More
Stainless Steel flake, -325 mesh, 0.4-1.2 micron thick, Type 316-L, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 013787.30
Code : NEW
Additional Details : Weight : 0.25000kg
Description
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel flake, Type 316-L | |
MFCD00148499 | |
Insoluble in water. | |
0.4 to 1.2 Micron thick |
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% | |
250 g | |
−325 Mesh | |
Flake |
RUO – Research Use Only