missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Stainless Steel disc, 152mm (6.0 in.) dia. x 12.7mm (0.5 in.) thick, Type 304

Brand:  Alfa Aesar™ 045183.KS

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 1 ea.

Product Code. 15476805

  • £94.00 / Each


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Specifications

Fe:Cr:Ni; 70:19:11 wt%
1 ea.
Odorless
Stainless Steel disc, Type 304
152mm (6.0 in.) dia. x 12.7mm (0.5 in.) thick
MFCD00148499
Videos
SDS