missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Stainless Steel Conductivity Probes

Brand:  Mettler Toledo™ 51344026

Code : NEW

Additional Details : Weight : 0.43000kg

 View more versions of this product

Product Code. 12669225

  • £515.00 / Each


Estimated Shipment
16-03-2021
Add to basket

Description

Description

Specifications

Specifications

For Measurements of Ultra-Pure Water
0.105 cm-1
Mini-DIN
Stainless Steel
Inlab741
0°C to 100°C
Conductivity Probe
120 mm
0.001 to 500 μS/cm
Videos
SDS
Documents