missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Stainless Steel Comfort Rack

Brand:  Tenak™ TE25124

Additional Details : Weight : 1.26000kg

 View more versions of this product

Product Code. 12405950

  • £116.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

140mm
Chest freezer
Boxes
Stainless Steel
1 Pack
Rack, Comfort
140 x 536mm
536mm
Spring lock
4
140mm
Videos