Learn More
ST8SIA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564115
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KENIFTTPKY ASPGAPRMYM FHAGFRSQFA LKFLDPSFVP ITNSLTQELQ EKPSKWKF Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.
ST8SIA3 Polyclonal antibody specifically detects ST8SIA3 in Human samples. It is validated for Immunohistochemistry.Specifications
ST8SIA3 | |
Polyclonal | |
Unconjugated | |
St8sia3 | |
AI847333; Alpha-2,8-sialyltransferase 8C; alpha-2,8-sialyltransferase III; GT3-synthase; sia-alpha-2,3-Gal-beta-1,4-GlcNAc-R:alpha 2,8-sialyltransferase; sialyltransferase 8 (alpha-2, 8-sialyltransferase) C; Sialyltransferase 8 (GT3 alpha 2,8-sialyltransferase) C; Sialyltransferase 8 (GT3 alpha 28-sialyltransferase) C; sialyltransferase 8 C; sialyltransferase 8C; sialyltransferase 8C (alpha2,3Galbeta1,4GlcNAcalpha 2,8-sialyltransferase); sialyltransferase St8Sia III; sialytransferase St8Sia III; SIAT8C; SIAT8-C; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 3; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3; St8sia3; ST8SiaIII | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51046 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43173 | |
St8sia3 | |
Recombinant protein corresponding to Human ST8SIA3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only