missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST2/IL-33R Rat anti-Mouse, Clone: 245714, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | ST2/IL-33R |
---|---|
Clone | 245714 |
Immunogen | S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse ST2/IL-33 R Ser27-Arg332 Accession # P14719 |
Gene ID (Entrez) | 9173 |
Gene Accession No. | P14719 |
Description
ST2/IL-33R Monoclonal specifically detects ST2/IL-33R in Mouse samples. It is validated for Western Blot.Specifications
ST2/IL-33R | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse ST2/IL-33 R Ser27-Arg332 Accession # P14719 | |
P14719 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Rat | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
245714 | |
9173 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse ST2/IL-33 R in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human ST2 and no cross-reactivity with recombinant mouse (rm) IL-1 R1 or rmIL-1 R2 is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only