missing translation for 'onlineSavingsMsg'
Learn More

ST 91, Tocris Bioscience™

α2 agonist, putative α2C agonist

Brand:  Tocris (Bio-Techne) 263850

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13451624

  • £467.46 / 50mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

α2-adrenoceptor agonist that does not cross the blood brain barrier. Displays ∽ 120-fold selectivity for α2 receptors over α1 receptors and acts predominantly at non-α2A-adrenoceptors which may be of the α2C subtype. Antinociceptive; increases tail-flick latencies in Sasco and Harlan rats.
Specifications

Chemical Identifiers

4749-61-5
253.774
n-2,6-diethylphenyl-4,5-dihydro-1h-imidazol-2-amine hydrochloride, benzenamine, 2,6-diethyl-n-2-imidazolidinylidene-, monohydrochloride
N-(2,6-diethylphenyl)-4,5-dihydro-1H-imidazol-2-amine;hydrochloride
C13H20ClN3
ZLRWFGBEDNTMEU-UHFFFAOYSA-N
185944
CCC1=C(C(=CC=C1)CC)NC2=NCCN2.Cl

Specifications

50mg
4749-61-5
n-2,6-diethylphenyl-4,5-dihydro-1h-imidazol-2-amine hydrochloride, benzenamine, 2,6-diethyl-n-2-imidazolidinylidene-, monohydrochloride
CCC1=C(C(=CC=C1)CC)NC2=NCCN2.Cl
253.774
253.77
ST 91
C13H20ClN3
ZLRWFGBEDNTMEU-UHFFFAOYSA-N
N-(2,6-diethylphenyl)-4,5-dihydro-1H-imidazol-2-amine;hydrochloride
185944
>99%
Videos
Documents