missing translation for 'onlineSavingsMsg'
Learn More

Somatostatin Recombinant Polyclonal Antibody (9HCLC), Invitrogen™

Rabbit Recombinant Polyclonal Antibody

Brand:  Invitrogen 710968

Additional Details : Weight : 0.01000kg

Product Code. 15652075

  • £396.00 / 100µg
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is predicted to react with Monkey, Goat, Sheep and Pig. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.

Somatostatin Recombinant Polyclonal antibody specifically detects Somatostatin in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.
Specifications

Specifications

Somatostatin
Recombinant Polyclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P61278, P60041
SST
Peptide corresponding to Human SST (aa 89-115)
100 μg
Primary
Human, Mouse
Antibody
IgG
Immunocytochemistry, Western Blot
9HCLC
Unconjugated
SST
Antrin; Growth hormone release-inhibiting factor; Neuronostatin; NST; preprosomatostatin; prepro-somatostatin; SMST; SOM; Somatostatin; somatostatin 28; somatostatin-14; Somatostatin-28; somatotropin release-inhibiting factor; SRIF; SS; SS-14; SS-28; Sst; SST-14
Rabbit
Protein A
RUO
20604, 6750
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers