missing translation for 'onlineSavingsMsg'
Learn More

SSR 146977 hydrochloride, Tocris Bioscience™

Potent and selective NK3 antagonist

Brand:  Tocris (Bio-Techne) 329750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13452734

  • £773.22 / 50mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective NK3 receptor antagonist (Ki values are 0.26 and 19.3 nM in CHO cells expressing the human NK3 and NK2 receptor respectively). Also inhibits senktide -induced inositol monophosphate formation and intracellular calcium mobilization (IC50 = 10 nM).
Specifications

Chemical Identifiers

264618-38-4
658.105
ssr 146977 hydrochloride, ssr 146977 hydrochloride/ssr-146977, n1-1-3-3r-1-benzoyl-3-3-3,4-dichlorophenyl-3-piperidinyl propyl-4-phenyl-piperidinyl-n,n-dimethylurea hydrochloride
3-[1-[3-[(3R)-1-benzoyl-3-(3,4-dichlorophenyl)piperidin-3-yl]propyl]-4-phenylpiperidin-4-yl]-1,1-dimethylurea;hydrochloride
C35H43Cl3N4O2
HYPKKQPFHSNZBY-GXUZKUJRSA-N
90488827
CN(C)C(=O)NC1(CCN(CC1)CCCC2(CCCN(C2)C(=O)C3=CC=CC=C3)C4=CC(=C(C=C4)Cl)Cl)C5=CC=CC=C5.Cl

Specifications

50mg
264618-38-4
ssr 146977 hydrochloride, ssr 146977 hydrochloride/ssr-146977, n1-1-3-3r-1-benzoyl-3-3-3,4-dichlorophenyl-3-piperidinyl propyl-4-phenyl-piperidinyl-n,n-dimethylurea hydrochloride
CN(C)C(=O)NC1(CCN(CC1)CCCC2(CCCN(C2)C(=O)C3=CC=CC=C3)C4=CC(=C(C=C4)Cl)Cl)C5=CC=CC=C5.Cl
658.105
658.11
SSR 146977 hydrochloride
C35H43Cl3N4O2
HYPKKQPFHSNZBY-GXUZKUJRSA-N
3-[1-[3-[(3R)-1-benzoyl-3-(3,4-dichlorophenyl)piperidin-3-yl]propyl]-4-phenylpiperidin-4-yl]-1,1-dimethylurea;hydrochloride
90488827
>98%
Videos
Documents