Learn More
SSH3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554118
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MKQYECSLEQ ALRHVQELRP IARPNPGFLR QLQIYQGILT ASRQSHVWEQ KVGGVSPEEH PAPEVSTPFP PLPPEPE Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 86%.
SSH3 Polyclonal antibody specifically detects SSH3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
SSH3 | |
Polyclonal | |
Unconjugated | |
Ssh3 | |
BC028922; hSSH-3L; mSSH-3L; Protein phosphatase Slingshot homolog 3; slingshot 3; slingshot homolog 3; slingshot homolog 3 (Drosophila); slingshot protein phosphatase 3; slingshot-3L; slingshot-like 3; Ssh3; SSH-3; Ssh3l; SSH-3L; SSH-like protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54961 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TE77 | |
Ssh3 | |
Recombinant protein corresponding to Human SSH3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only