missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA4 Monoclonal Antibody (MC-813-70), Alexa Fluor™ 647, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen SSEA421
176.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00050kg
Description
SSEA4 Monoclonal antibody specifically detects SSEA4 in Human samples. It is validated for Flow Cytometry.Specifications
SSEA4 | |
Monoclonal | |
Alexa Fluor 647 | |
SSEA-4; stage-specific embryonic antigen 4 | |
Human SSEA4 antigen | |
500 μL | |
Primary | |
4° C | |
Liquid |
Flow Cytometry | |
MC-813-70 | |
PBS with 0.1% sodium azide | |
Mouse | |
Affinity chromatography | |
RUO | |
Human | |
Antibody | |
IgG3 |