missing translation for 'onlineSavingsMsg'
Learn More

SSEA4, PerCP-eFluor™ 710, clone: eBioMC-813-70 (MC-813-70), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-8843-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15569376

  • £119.00 / 25 tests

Estimated Shipment
26-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The eBioMC-813-70 monoclonal antibody reacts with a glycolipid carbohydrate epitope on Stage-specific embryonic antigen-4 (SSEA-4) protein. Expression is found on human teratocarcinoma stem cells (EC), human embryonic germ cells (EG), human induced pluripotent stem (iPS) cells, and human embryonic stem cells (ES) with a decrease in expression upon differentiation. Recently expression has also been found on bone marrow mesenchymal stem cells.The eBioMC-813-70 antibody has been reported to crossreact to mouse, rabbit, canine, chicken, and Old and New World monkeys.Applications Reported: This eBioMC-813-70 (MC-813-70) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioMC-813-70 (MC-813-70) antibody has been pre-titrated and tested by flow cytometric analysis of the 2102Ep cell line. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor™ 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor™ 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

SSEA4
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
stage-specific embryonic antigen 4, SSEA-4
IgG3
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
Canine, Chicken, Human, Murine, Non-human Primate, Rabbit
Flow Cytometry
eBioMC-813-70 (MC-813-70)
PerCP-eFluor 710
Liquid
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Videos
Documents

For Research Use Only.