Learn More
SSEA4 Monoclonal Antibody (eBioMC-813-70 (MC-813-70)), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 46-8843-42
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The eBioMC-813-70 monoclonal antibody reacts with a glycolipid carbohydrate epitope on Stage-specific embryonic antigen-4 (SSEA-4) protein. Expression is found on human teratocarcinoma stem cells (EC), human embryonic germ cells (EG), human induced pluripotent stem (iPS) cells, and human embryonic stem cells (ES) with a decrease in expression upon differentiation. Recently expression has also been found on bone marrow mesenchymal stem cells. The eBioMC-813-70 antibody has been reported to crossreact to mouse, rabbit, canine, chicken, and Old and New World monkeys. Applications Reported: This eBioMC-813-70 (MC-813-70) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioMC-813-70 (MC-813-70) antibody has been pre-titrated and tested by flow cytometric analysis of the 2102Ep cell line. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor™ 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome.
SSEA4 Monoclonal antibody specifically detects SSEA4 in Canine, Chicken, Human, Mouse, Non-human Primate, Rabbit samples. It is validated for Flow Cytometry.Specifications
SSEA4 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Mouse | |
100 Tests | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
eBioMC-813-70 (MC-813-70) | |
PerCP-eFluor 710 | |
SSEA-4; stage-specific embryonic antigen 4 | |
Affinity chromatography | |
RUO | |
Canine, Chicken, Human, Mouse, Non-human Primate, Rabbit | |
Antibody | |
IgG3 |
For Research Use Only.