missing translation for 'onlineSavingsMsg'
Learn More

SSEA4, eFluor 570, clone: eBioMC-813-70 (MC-813-70), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 41-8843-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15331080

  • £174.38 / 100µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The eBioMC-813-70 monoclonal antibody reacts with a glycolipid carbohydrate epitope on Stage-specific embryonic antigen-4 (SSEA-4) protein. Expression is found on human teratocarcinoma stem cells (EC), human embryonic germ cells (EG), human induced pluripotent stem (iPS) cells, and human embryonic stem cells (ES) with a decrease in expression upon differentiation. Recently expression has also been found on bone marrow mesenchymal stem cells.The eBioMC-813-70 antibody has been reported to crossreact to mouse, rabbit, canine, chicken, and Old and New World monkeys.Applications Reported: This eBioMC-813-70 (MC-813-70) antibody has been reported for use in immunocytochemistry. This product has not been validated for flow cytometric analysis.Applications Tested: This eBioMC-813-70 (MC-813-70) antibody has been tested by immunocytochemistry on fixed NTERA-2 and 2102Ep cells and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filter Recommendation: When using this eFluor® 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor® 555 or TRITC filter is acceptable.Excitation: 555 nm; Emission: 570 nm
Specifications

Specifications

SSEA-4
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
stage-specific embryonic antigen 4, SSEA-4
IgG3
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
CanineþChickenþHumanþMurineþNon-human PrimateþRabbit
Immunocytochemistry
eBioMC-813-70 (MC-813-70)
eFluor 570
Conjugated
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Videos

For Research Use Only.