Learn More
SSEA4 Monoclonal Antibody (eBioMC-813-70 (MC-813-70)), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 53-8843-42
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The eBioMC-813-70 monoclonal antibody reacts with a glycolipid carbohydrate epitope on Stage-specific embryonic antigen-4 (SSEA-4) protein. Expression is found on human teratocarcinoma stem cells (EC), human embryonic germ cells (EG), human induced pluripotent stem (iPS) cells, and human embryonic stem cells (ES) with a decrease in expression upon differentiation. Recently expression has also been found on bone marrow mesenchymal stem cells. The eBioMC-813-70 antibody has been reported to crossreact to mouse, rabbit, canine, chicken, and Old and New World monkeys. Applications Reported: This eBioMC-813-70 (MC-813-70) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioMC-813-70 (MC-813-70) antibody has been pre-titrated and tested by flow cytometric analysis of the 2102Ep cell line. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
SSEA4 Monoclonal antibody specifically detects SSEA4 in Canine, Chicken, Human, Mouse, Non-human Primate, Rabbit samples. It is validated for Flow Cytometry.Specifications
SSEA4 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Mouse | |
100 Tests | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
eBioMC-813-70 (MC-813-70) | |
Alexa Fluor 488 | |
SSEA-4; stage-specific embryonic antigen 4 | |
Affinity chromatography | |
RUO | |
Canine, Chicken, Human, Mouse, Non-human Primate, Rabbit | |
Antibody | |
IgG3 |
For Research Use Only.