missing translation for 'onlineSavingsMsg'
Learn More

SSEA3, Biotin, clone: eBioMC-631 (MC-631), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-8833-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15300080

  • £87.94 / 25µg


Estimated Shipment date
from Supplier 28-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The eBioMC-631 (MC-631) monoclonal antibody reacts with the carbohydrate moiety galactosylgloboside on Stage-specific embryonic antigen-3 (SSEA-3). Expression is restricted to the surface of human teratocarcinoma stem cells (EC), human embryonic germ cells (EG) and human embryonic stem cells (ES) and down regulated following differentiation.Applications Reported: This eBioMC-631 (MC-631) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioMC-631 (MC-631) antibody has been tested by flow cytometric analysis of the 2102Ep cell line. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

SSEA-3
eBioMC-631 (MC-631)
Biotin
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
stage-specific embryonic antigen-3
IgM
RUO
Primary
Murine
Videos