Learn More
SSEA3 Monoclonal Antibody (eBioMC-631 (MC-631)), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£132.00 - £263.00
Specifications
Antigen | SSEA3 |
---|---|
Clone | eBioMC-631 (MC-631) |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15550257
|
Affymetrix eBioscience
53-8833-41 |
25 Tests |
£132.00
25 tests |
Estimated Shipment: 01-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15560257
|
Affymetrix eBioscience
53-8833-42 |
100 Tests |
£263.00
100 tests |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The eBioMC-631 (MC-631) monoclonal antibody reacts with the carbohydrate moiety galactosylgloboside on Stage-specific embryonic antigen-3 (SSEA-3). Expression is restricted to the surface of human teratocarcinoma stem cells (EC), human embryonic germ cells (EG) and human embryonic stem cells (ES) and down regulated following differentiation. Applications Reported: This eBioMC-631 (MC-631) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioMC-631 (MC-631) antibody has been pre-titrated and tested by flow cytometric analysis of the 2102Ep cell line. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
SSEA3 Monoclonal antibody specifically detects SSEA3 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
SSEA3 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
IgM | |
Affinity chromatography | |
Antibody |
eBioMC-631 (MC-631) | |
Flow Cytometry | |
Alexa Fluor 488 | |
Rat | |
Human, Mouse | |
SSEA-3; stage-specific embryonic antigen 3 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.