missing translation for 'onlineSavingsMsg'
Learn More

SSEA3, Alexa Fluor 488, clone: eBioMC-631 (MC-631), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-8833-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15560257

  • £199.07 / 100 tests


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Description: The eBioMC-631 (MC-631) monoclonal antibody reacts with the carbohydrate moiety galactosylgloboside on Stage-specific embryonic antigen-3 (SSEA-3). Expression is restricted to the surface of human teratocarcinoma stem cells (EC), human embryonic germ cells (EG) and human embryonic stem cells (ES) and down regulated following differentiation.Applications Reported: This eBioMC-631 (MC-631) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioMC-631 (MC-631) antibody has been pre-titrated and tested by flow cytometric analysis of the 2102Ep cell line. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

SSEA-3
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
stage-specific embryonic antigen 3, SSEA-3
IgM
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
HumanĂ¾Murine
Flow Cytometry
eBioMC-631 (MC-631)
Alexa Fluor 488
Conjugated
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Videos

For Research Use Only.