Learn More
SSEA1 Monoclonal Antibody (eBioMC-480 (MC-480)), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 46-8813-42
197.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.23750kg
Description
CD15 (Lewis X, Le(x); stage specific embryonic antigen-1, SSEA-1) is a trisacharide determinant (3-fucosyl-N-acetyllactosamine) expressed on several glycolipids, glycoproteins and proteoglycans of various cell types, e.g. granulocytes, mast cells, monocytes, macrophages, cells of gastric mucosa, nervous system or various tumour cells. There are several variants of Lewis x, such as sialyl-Lewis x or sulphated Lewis x. Cells with high surface expression of Le(x) antigen exhibit strong self-aggregation, based on calcium-dependent Le(x)-Le(x) interaction. This process is involved for example in embryo compaction or in autoaggregation of teratocarcinoma cells. Sialyl-Le(x) and its isomer sialyl-Le(a) are ligands of selectins. CD15 expression has been extensively used to confirm diagnosis of Hodgkin's disease.
SSEA1 Monoclonal antibody specifically detects SSEA1 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
SSEA1 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P22083, Q11127 | |
FUT4 | |
Affinity chromatography | |
RUO | |
14345, 2526 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
eBioMC-480 (MC-480) | |
PerCP-eFluor 710 | |
FUT4 | |
stage-specific embryonic antigen 1, CD15, SSEA-1, SSEA1, ELFT, FCT3A, FUC-TIV, FUTIV, LeX | |
Mouse | |
100 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgM |
For Research Use Only.