missing translation for 'onlineSavingsMsg'
Learn More

SSEA1, PE, clone: eBioMC-480 (MC-480), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-8813-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15537136

  • £81.20 / 25 tests


EU Stock 1
Add to basket

Description

Description

Description: The eBioMC-480 (MC-480) antibody reacts with the stage-specific embryonic antigen-1 (SSEA-1), a carbohydrate epitope expressed upon the surface of early mouse embryos, murine embryonal carcinoma cells (EC), murine embryonic stem cells (ES) and murine and human germ cells (EG). No immunoreactivity is evident with undifferentiated human EC and ES cells. Differentiation of human EC results in an increase in SSEA-1 expression, while in the mouse expression is diminished. Expression of the carbohydrate moiety is also found on mature human granulocytes (on CD15) and some monocytes. SSEA-1 is associated with cell adhesion, migration and differentiation.Applications Reported: This eBioMC-480 (MC-480) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioMC-480 (MC-480) antibody has been pre-titrated and tested by flow cytometric analysis of the F9 cell line. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

SSEA-1
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
stage-specific embryonic antigen 1, CD15, SSEA-1, SSEA1, ELFT, FCT3A, FUC-TIV, FUTIV, LeX
IgM
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
Murine
Flow Cytometry
eBioMC-480 (MC-480)
PE
Conjugated
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Videos

For Research Use Only.