Learn More
SRRM2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559559
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RSRSSSPVTE LASRSPIRQD RGEFSASPML KSGMSPEQSR FQSDSSSYPT VDSNSLLGQS RLETAESKEK MALPPQEDAT ASPPRQKDKF SPFPVQDRPE SSLVFK Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 60%.
SRRM2 Polyclonal antibody specifically detects SRRM2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
SRRM2 | |
Polyclonal | |
Unconjugated | |
SRRM2 | |
300 kDa nuclear matrix antigen; 300-KD; 5033413A03Rik; AA410130; Cwc21; CWF21; HSPC075; Kiaa0324; mKIAA0324; RNA binding protein; ser/Arg-related nuclear matrix protein of 300 kDa; serine/arginine repetitive matrix 2; serine/arginine repetitive matrix protein 2; Serine/arginine-rich splicing factor-related nuclear matrix protein of 300 kDa; splicing coactivator subunit SRm300; SRL300; SRm300; SR-related nuclear matrix protein of 300 kDa; SRRM2; taxREB803; Tax-responsive enhancer element-binding protein 803; testicular secretory protein Li 53 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23524 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UQ35 | |
SRRM2 | |
Recombinant protein corresponding to Human SRRM2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only