missing translation for 'onlineSavingsMsg'
Learn More

SR 59230A hydrochloride, Tocris Bioscience™

Potent and selective β3 antagonist

Brand:  Tocris (Bio-Techne) 151110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13459583

  • £175.56 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Potent and selective β3 adrenoceptor antagonist (IC50 values are 40, 408 and 648 nM for β3, β1 and β2 receptors respectively). Orally active in vivo. Also available as part of the β-Adrenoceptor Antagonist Tocriset™ .
Specifications

Chemical Identifiers

1135278-41-9
361.91
sr 59230a hydrochloride, 1-2-ethylphenoxy-3-1s-1,2,3,4-tetrahydro-1-naphthalenyl amino-2s-2-propanol hydrochloride, 2s-1-2-ethylphenoxy-3-1s-1,2,3,4-tetrahydronaphthalen-1-ylamino propan-2-ol hydrochloride
(2S)-1-(2-ethylphenoxy)-3-[[(1S)-1,2,3,4-tetrahydronaphthalen-1-yl]amino]propan-2-ol;hydrochloride
C21H28ClNO2
SHUCXUIOEAAJJL-MKSBGGEFSA-N
24978529
CCC1=CC=CC=C1OCC(CNC2CCCC3=CC=CC=C23)O.Cl

Specifications

SR 59230A hydrochloride
1135278-41-9
sr 59230a hydrochloride, 1-2-ethylphenoxy-3-1s-1,2,3,4-tetrahydro-1-naphthalenyl amino-2s-2-propanol hydrochloride, 2s-1-2-ethylphenoxy-3-1s-1,2,3,4-tetrahydronaphthalen-1-ylamino propan-2-ol hydrochloride
CCC1=CC=CC=C1OCC(CNC2CCCC3=CC=CC=C23)O.Cl
361.91
361.91
10mg
C21H28ClNO2
SHUCXUIOEAAJJL-MKSBGGEFSA-N
(2S)-1-(2-ethylphenoxy)-3-[[(1S)-1,2,3,4-tetrahydronaphthalen-1-yl]amino]propan-2-ol;hydrochloride
24978529
>98%
Videos