Learn More
SR 57227 hydrochloride, Tocris Bioscience™
Potent, selective 5-HT3 agonist
Brand:  Tocris (Bio-Techne) 120510
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13458863
  • £128.25 / 10mg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
A high affinity, selective 5-HT3 receptor agonist, active in vivo and CNS penetrant.
Specifications
Chemical Identifiers
77145-61-0
248.151
sr 57227 hydrochloride, unii-ow45b79uzd, sr 57227a hydrochloride, 4-amino-1-6-chloro-2-pyridyl-piperidine hydrochloride, ow45b79uzd, 1-6-chloropyridin-2-yl piperidin-4-amine hydrochloride, 4-amino-1-6-chloro-2-pyridyl piperidine hydrochloride, 1-6-chloro-2-pyridinyl-4-piperidinamine monohydrochloride, 4-amino-1-6-chloro-2-pyridyl piperidine monohydrochloride, 4-piperidinamine, 1-6-chloro-2-pyridinyl-, monohydrochloride
1-(6-chloropyridin-2-yl)piperidin-4-amine;hydrochloride
C10H15Cl2N3
FUMINTAAUJUVMP-UHFFFAOYSA-N
131746
C1CN(CCC1N)C2=NC(=CC=C2)Cl.Cl
Specifications
SR 57227 hydrochloride
77145-61-0
sr 57227 hydrochloride, unii-ow45b79uzd, sr 57227a hydrochloride, 4-amino-1-6-chloro-2-pyridyl-piperidine hydrochloride, ow45b79uzd, 1-6-chloropyridin-2-yl piperidin-4-amine hydrochloride, 4-amino-1-6-chloro-2-pyridyl piperidine hydrochloride, 1-6-chloro-2-pyridinyl-4-piperidinamine monohydrochloride, 4-amino-1-6-chloro-2-pyridyl piperidine monohydrochloride, 4-piperidinamine, 1-6-chloro-2-pyridinyl-, monohydrochloride
C1CN(CCC1N)C2=NC(=CC=C2)Cl.Cl
248.151
248.15
10mg
C10H15Cl2N3
FUMINTAAUJUVMP-UHFFFAOYSA-N
1-(6-chloropyridin-2-yl)piperidin-4-amine;hydrochloride
131746
>99%
Videos