Learn More
Tocris Bioscience™ SR 48692
Selective non-peptide NTS1 antagonist
Brand:  Tocris Bioscience™ 3721/50
Code : Z1
Additional Details : Weight : 0.00050kg
 View more versions of this product
Product Code. 13463484
  • £863.96 / 50mg
Estimated Shipment
04-04-2019
Add to basket
Description
Description
Neurotensin antagonist; selective for NTS1 over NTS2 (apparent affinity, Ke, is 36 nM for NTS1). Competitively inhibits binding of [125I]-neurotensin to HT29 and N1E115 cell membranes (IC50 values are 15.3 and 20.4 nM respectively). Orally bioavailable.
Specifications
Specifications
SR 48692
>98
50mg
SR 48692
587.07