missing translation for 'onlineSavingsMsg'
Learn More

SR 2640 hydrochloride, Tocris Bioscience™

Potent, selective LTD4/LTE4 receptor antagonist

£149.94 - £573.30

Chemical Identifiers

CAS 146662-42-2
Molecular Formula C23H19ClN2O3
Molecular Weight (g/mol) 406.866
InChI Key KDSYWFCTUKABKE-UHFFFAOYSA-N
Synonym sr 2640 hydrochloride, 2-3-2-quinolinylmethoxy phenyl amino benzoic acid hydrochloride, 2-3-quinolin-2-ylmethoxy phenyl amino benzoic acid hydrochloride, 2-3-2-quinolinylmethoxy phenyl amino benzoic acidhydrochloride, benzoic acid,2-3-2-quinolinylmethoxy phenyl amino, 2-3-2-quinolinylmethoxy phenyl a??mino benzoic acid hydrochloride
PubChem CID 45073453
IUPAC Name 2-[3-(quinolin-2-ylmethoxy)anilino]benzoic acid;hydrochloride
SMILES C1=CC=C2C(=C1)C=CC(=N2)COC3=CC=CC(=C3)NC4=CC=CC=C4C(=O)O.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13430034 View Documents Tocris (Bio-Techne)
180410
10mg £149.94
10mg View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
13440034 View Documents Tocris (Bio-Techne)
180450
50mg £573.30
50mg View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potent and selective competitive leukotriene D4 and E4 receptor antagonist. Inhibits LTD4- but not histamine-induced guinea pig ileum and trachea contraction (pA2 = 8.7). Also inhibits LTD4-attenuation of human PMN chemotaxis. Orally active in vivo.
Specifications

Chemical Identifiers

146662-42-2
406.866
sr 2640 hydrochloride, 2-3-2-quinolinylmethoxy phenyl amino benzoic acid hydrochloride, 2-3-quinolin-2-ylmethoxy phenyl amino benzoic acid hydrochloride, 2-3-2-quinolinylmethoxy phenyl amino benzoic acidhydrochloride, benzoic acid,2-3-2-quinolinylmethoxy phenyl amino, 2-3-2-quinolinylmethoxy phenyl a??mino benzoic acid hydrochloride
2-[3-(quinolin-2-ylmethoxy)anilino]benzoic acid;hydrochloride
C23H19ClN2O3
KDSYWFCTUKABKE-UHFFFAOYSA-N
45073453
C1=CC=C2C(=C1)C=CC(=N2)COC3=CC=CC(=C3)NC4=CC=CC=C4C(=O)O.Cl

Specifications

10mg
146662-42-2
sr 2640 hydrochloride, 2-3-2-quinolinylmethoxy phenyl amino benzoic acid hydrochloride, 2-3-quinolin-2-ylmethoxy phenyl amino benzoic acid hydrochloride, 2-3-2-quinolinylmethoxy phenyl amino benzoic acidhydrochloride, benzoic acid,2-3-2-quinolinylmethoxy phenyl amino, 2-3-2-quinolinylmethoxy phenyl a??mino benzoic acid hydrochloride
C1=CC=C2C(=C1)C=CC(=N2)COC3=CC=CC(=C3)NC4=CC=CC=C4C(=O)O.Cl
406.866
406.87
SR 2640 hydrochloride
C23H19ClN2O3
KDSYWFCTUKABKE-UHFFFAOYSA-N
2-[3-(quinolin-2-ylmethoxy)anilino]benzoic acid;hydrochloride
45073453
>99%