missing translation for 'onlineSavingsMsg'
Learn More

SR 2640 hydrochloride, Tocris Bioscience™

Potent, selective LTD4/LTE4 receptor antagonist

Brand:  Tocris (Bio-Techne) 180450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13440034

  • £573.30 / 50mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective competitive leukotriene D4 and E4 receptor antagonist. Inhibits LTD4- but not histamine-induced guinea pig ileum and trachea contraction (pA2 = 8.7). Also inhibits LTD4-attenuation of human PMN chemotaxis. Orally active in vivo.
Specifications

Chemical Identifiers

146662-42-2
406.866
sr 2640 hydrochloride, 2-3-2-quinolinylmethoxy phenyl amino benzoic acid hydrochloride, 2-3-quinolin-2-ylmethoxy phenyl amino benzoic acid hydrochloride, 2-3-2-quinolinylmethoxy phenyl amino benzoic acidhydrochloride, benzoic acid,2-3-2-quinolinylmethoxy phenyl amino, 2-3-2-quinolinylmethoxy phenyl a??mino benzoic acid hydrochloride
2-[3-(quinolin-2-ylmethoxy)anilino]benzoic acid;hydrochloride
C23H19ClN2O3
KDSYWFCTUKABKE-UHFFFAOYSA-N
45073453
C1=CC=C2C(=C1)C=CC(=N2)COC3=CC=CC(=C3)NC4=CC=CC=C4C(=O)O.Cl

Specifications

50mg
146662-42-2
sr 2640 hydrochloride, 2-3-2-quinolinylmethoxy phenyl amino benzoic acid hydrochloride, 2-3-quinolin-2-ylmethoxy phenyl amino benzoic acid hydrochloride, 2-3-2-quinolinylmethoxy phenyl amino benzoic acidhydrochloride, benzoic acid,2-3-2-quinolinylmethoxy phenyl amino, 2-3-2-quinolinylmethoxy phenyl a??mino benzoic acid hydrochloride
C1=CC=C2C(=C1)C=CC(=N2)COC3=CC=CC(=C3)NC4=CC=CC=C4C(=O)O.Cl
406.866
406.87
SR 2640 hydrochloride
C23H19ClN2O3
KDSYWFCTUKABKE-UHFFFAOYSA-N
2-[3-(quinolin-2-ylmethoxy)anilino]benzoic acid;hydrochloride
45073453
>99%
Videos
Documents